Learn More
R&D Systems™ Cultrex Basement Membrane Extract, Type 2
Applications: Bioactivity
Manufacturer:  R&D Systems™ 353201002
 View more versions of this product
Catalog No. 35-320-1002
Add to cart
This item is not returnable. View return policy
Description
Description
Functional assay: Tube formation assay - BME promotes formation of capillary-like structures by human (HBMVEC; HUVEC) or mouse (SVEC4-10) endothelial cells. Gelling assay: BME gels in less than 30 minutes at 37degreesC, and maintains the gelled form in culture medium for a minimum of 14 days at 37degreesC.
Specifications
Specifications
12-18mg/mL
2 x 5mL
Murine Engelbreth-Holm-Swarm (EHS) tumor.
Product is stable for a minimum of 3 months from date of shipment when stored at -20° C in a manual defrost freezer. For optimal stability, store at -80° C. Avoid freeze-thaw cycles.
12 - 18mg/mL
Bioactivity