Learn More
R&D Systems™ Cultrex RGF Basement Membrane Extract, Type 2
For Robust Organoid Cultures - Reduced Growth Factor Basement Membrane Extract
$68.90 - $517.36
Specifications
Concentration | 12 to 18 mg/mL |
---|---|
For Use With (Application) | Bioactivity |
Source | Murine Engelbreth-Holm-Swarm (EHS) tumor. |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
35-330-0102
|
R&D Systems™
353300102 |
1 mL |
Each for $68.90
|
|
|||||
35-330-0502
|
R&D Systems™
353300502 |
5 mL |
Each for $322.48
|
|
|||||
35-330-1002
|
R&D Systems™
353301002 |
2 x 5 mL |
Each for $517.36
|
|
|||||
Description
Functional Assay - Tube formation assay: BME promotes formation of capillary-like structures by human (HBMVEC; HUVEC) or mouse (SVEC4-10) endothelial cells. Gelling Assay - BME gels in less than 30 minutes at 37°C, and maintains the gelled form in culture medium for a minimum of 14 days at 37°C.Specifications
12 to 18 mg/mL | |
Bioactivity | |
Murine Engelbreth-Holm-Swarm (EHS) tumor. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.