Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Cultrex Basement Membrane Extract, Type 3, Pathclear Available on GSA/VA Contract for Federal Government customers only.

Applications: Bioactivity

$43.00 - $249.00

Specifications

Concentration 12-18mg/mL
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
36-320-0102
View Documents
R&D Systems™
363200102
1mL
Each for $43.00
Add to cart
 
36-320-1002 Available on GSA/VA Contract for Federal Government customers only.
View Documents
R&D Systems™
363201002
2x5mL
Each for $249.00
Add to cart
 
36-320-0502
View Documents
R&D Systems™
363200502
5mL
Each for $175.40
Add to cart
 
Description

Description

Basement membranes are continuous sheets of specialized extracellular matrix that form an interface between endothelial, epithelial, muscle, or neuronal cells and their adjacent stroma. Basement membranes are degraded and regenerated during development and wound healing. They not only support cells and cell layers, but they also play an essential role in tissue organization that affects cell adhesion, migration, proliferation, and differentiation. Basement membranes provide major barriers to invasion by metastatic tumor cells.

The major components of BME include laminin, collagen IV, entactin, and heparin sulfate proteoglycan
BME can be used in a multiple applications, under a variety of cell culture conditions, for maintaining growth or promoting differentiation of primary endothelial, epithelial, smooth muscle and stem cells
BME can also be utilized in cell attachment, neurite outgrowth, angiogenesis, in vitro cell invasion and in vivo tumorigenicity assays
Higher tensile strength than standard BME
DMEM without phenol red storage buffer
Sterility tested

Applications:

solid tumors

Specifications

Specifications

12-18mg/mL
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit