Learn More
R&D Systems™ Directed In Vivo Angiogenesis Assay Starter Kit
Designed to assess angiogenesis in an in vivo system
Manufacturer:  R&D Systems™ 3450048SK
Catalog No. 3450048SK
Add to cart
Includes: Angioreactors; Growth Factor Reduced PathClear™ BME; 10X Wash Buffer; CellSperse™; 200X FITC-Lectin; 25X FITC-Lectin Diluent; Heparin Solution; FGF-2 (1.8ug)/VEGF, 600ng
This item is not returnable. View return policy
Description
Description
During the course of the assay, implant grade silicone cylinders closed at one end, called angioreactors, are filled with basement membrane extract (BME)
These angioreactors are then implanted subcutaneously in the dorsal flanks of nude mice
If filled w
Specifications
Specifications
Each