Learn More
Tocris Bioscience™ DSR 6434
Potent TLR7 agonist
$232.00 - $964.00
Specifications
Product Type Bioactive Small Molecule
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
480910
View Documents Promotion Details
Tocris Bioscience™
4809/10
10 mg
Each for $232.00
Add to cart
480950
View Documents Promotion Details
Tocris Bioscience™
4809/50
50 mg
Each for $964.00
Add to cart
Description
Description
Potent TLR7 agonist (EC50 = 7.2 nM). Activates several immune effector cells in vitro. Suppresses metastasis in vivo and enhances efficacy of radiation therapy in a model of colorectal carcinoma.
Specifications
Specifications
Bioactive Small Molecule
Safety and Handling
Safety and Handling
Recommended Storage : Store at -20°C
SDS
Product Certifications
Research Use Only. Not For Human or Veterinary Use