Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Equine IL-1ra/IL-1F3 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
$490.68
Specifications
Molecular Weight (g/mol) | 17.5 kDa |
---|---|
Name | IL-1ra/IL-1F3 |
Quantity | 50μg |
Source | E. coli-derived equine IL-1ra/IL-1F3 protein His26-Gln177,with an N-terminal Met |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
2466RA050
|
R&D Systems™
2466RA050 |
Lyophilized from a 0.2μm filtered solution in PBS and DTT with BSA as a carrier protein. |
Each for $490.68
|
|
|||||
2466RA050CF
|
R&D Systems™
2466RA050/CF |
Lyophilized from a 0.2μm filtered solution in PBS and DTT. |
Each for $490.68
|
|
|||||
Description
The Recombinant Equine IL-1ra/IL-1F3 Protein is derived from E. coli. The Recombinant Equine IL-1ra/IL-1F3 Protein has been validated for the following applications: Bioactivity.Specifications
17.5 kDa | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
IL-1ra/IL-1F3 | |
E. coli-derived equine IL-1ra/IL-1F3 protein His26-Gln177,with an N-terminal Met | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title