Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Feline IL-12 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
$531.28
Specifications
Gene ID (Entrez) | 493741 |
---|---|
Molecular Weight (g/mol) | 23.7 kDa (p35), 36.2 kDa (p40) |
Name | IL-12 |
Quantity | 25μg |
Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived feline IL-12 protein, (N-terminus) Feline IL-12 p40 (Ile23-Ser329) (Glu167Gly) Accession &Num; O02744 10-His tag Feline IL-12 p35 (Arg26-Ser222) (Ala221Ser) Accession &Num; O02743 10-His tag (C-terminus) |
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
1954FL025CF
|
R&D Systems™
1954FL025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
Each for $531.28
|
|
|||||
1954FL025
|
R&D Systems™
1954FL025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
Each for $531.28
|
|
|||||
Description
The Recombinant Feline IL-12 Protein is derived from Sf 21 (baculovirus). The Recombinant Feline IL-12 Protein has been validated for the following applications: Bioactivity.Specifications
493741 | |
IL-12 | |
Spodoptera frugiperda,Sf 21 (baculovirus)-derived feline IL-12 protein, (N-terminus) Feline IL-12 p40 (Ile23-Ser329) (Glu167Gly) Accession &Num; O02744 10-His tag Feline IL-12 p35 (Arg26-Ser222) (Ala221Ser) Accession &Num; O02743 10-His tag (C-terminus) | |
Disulfide-linked heterodimer | |
Recombinant |
23.7 kDa (p35), 36.2 kDa (p40) | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title