Learn More
R&D Systems™ Recombinant Feline IL-2 (Cys146Ser) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Supplier: R&D Systems™ 1890FL020
This item is not returnable.
View return policy
Description
The Recombinant Feline IL-2 (Cys146Ser) Protein is derived from E. coli. The Recombinant Feline IL-2 (Cys146Ser) Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in Sodium Acetate with BSA as a carrier protein. | |
15.6 kDa | |
20μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
751114 | |
IL-2 (Cys146Ser) | |
E. coli-derived feline IL-2 protein Ala21-Thr154 (Cys146Ser),with and without an N-terminal Met | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |