Learn More
R&D Systems Feline IL-5 Recombinant Protein
Manufacturer:  R&D Systems 1795FL010
 View more versions of this product
Catalog No. 1795FL010
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
14.6 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Feline
Unlabeled
493803
IL-5
10μg
Disulfide-linked homodimer
Yes