Learn More
Tocris Bioscience™ GlyH 101
Reversible, voltage-dependent CFTR chloride channel blocker
$211.00 - $883.00
Specifications
Product Type Bioactive Small Molecule
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
548510
View Documents
Tocris Bioscience™
5485/10
10 mg
Each for $211.00
Add to cart
548550
View Documents
Tocris Bioscience™
5485/50
50 mg
Each for $883.00
Add to cart
Description
Description
Reversible, voltage-dependent CFTR chloride channel blocker (Ki = 4.3 μM). Inhibits forskolin -induced hyperpolarization in nasal potential differences and inhibits cholera toxin-induced intestinal fluid secretion in mice.
Specifications
Specifications
Bioactive Small Molecule
Safety and Handling
Safety and Handling
Recommended Storage : Store at -20°C
SDS
Product Certifications
Research Use Only. Not For Human or Veterinary Use