Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ GSK LSD 1 Dihydrochloride
Potent and selective LSD1 inhibitor
$248.24 - $1038.20
Specifications
Chemical Name or Material | GSK LSD 1 dihydrochloride |
---|---|
CAS | 2102933-95-7 |
Synonym | rel-N-[(1S,2R)-2-Phenylcyclopropyl]-4-piperidinamine dihydrochloride |
Solubility | Soluble to 100mM in water and to 100mM in DMSO |
Molecular Formula | C14H20N2·2HCl |
Description
GSK LSD 1 dihydrochloride is a potent and selective lysine specific demethylase 1 (LSD1) inhibitor (IC50 = 16 nM). Exhibits >1000-fold selectivity for LSD1 over closely related enzymes including LSD2, MAO-1 and MAO-B. Inhibits cancer cell line growth in vitro.Specifications
GSK LSD 1 dihydrochloride | |
rel-N-[(1S,2R)-2-Phenylcyclopropyl]-4-piperidinamine dihydrochloride | |
C14H20N2·2HCl | |
Solid | |
>98% |
2102933-95-7 | |
Soluble to 100mM in water and to 100mM in DMSO | |
289.24 | |
Bioactive Small Molecule |
Safety and Handling
Recommended Storage : Store at 4°C
Research Use Only. Not For Human or Veterinary Use
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title