Learn More
R&D Systems™ Herring Sperm DNA
5 vials (1 mL/vial) of Herring Sperm DNA
Manufacturer:  R&D Systems™ 96055D
Catalog No. 96055D
Add to cart
This item is not returnable. View return policy
Description
Description
  • Fragment size range of 200 - 500bp
  • The format allows convenient addition to buffers to create a final recommended working concentration of 0.1mg/mL
  • Quality control testing includes concentration determination, fragment sizing by gel electrophoresis, A260/A280 ultraviolet absorbance ratio, and protein content
Specifications
Specifications
Herring Sperm DNA
5 x 1 mL
Store at -20° C in a manual defrost freezer. For long-term storage, freeze in working aliquots at -80° C. Avoid freeze-thaw cycles.