Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Angiopoietin-4 Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

$579.00

Specifications

Gene ID (Entrez) 51378
Molecular Weight (g/mol) 57 kDa (monomer)
Name Angiopoietin-4
Quantity 25μg
Source Mouse myeloma cell line,NS0-derived human Angiopoietin-4 protein, (N-terminus) Human Ang-4 (Gln23-Ile503) or (Gln24-Ile503) Accession &Num; Q9Y264 IEGR GGGSGGGSGGGS 10-His tag (C-terminus)
View More Specs

Products 2
Catalog Number Mfr. No. Formulation Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Price Quantity & Availability  
964AN025
View Documents
R&D Systems™
964AN025
Lyophilized from a 0.2μm filtered solution in Tris-Citrate and NaCl with BSA as a carrier protein.
Each for $579.00
Add to Cart
 
964AN025CF
View Documents
R&D Systems™
964AN025/CF
Lyophilized from a 0.2μm filtered solution in Tris-Citrate and NaCl.
Each for $579.00
Add to Cart
 
Description

Description

The Recombinant Human Angiopoietin-4 Protein is derived from NS0. The Recombinant Human Angiopoietin-4 Protein has been validated for the following applications: Binding Activity.
Specifications

Specifications

51378
Angiopoietin-4
Mouse myeloma cell line,NS0-derived human Angiopoietin-4 protein, (N-terminus) Human Ang-4 (Gln23-Ile503) or (Gln24-Ile503) Accession &Num; Q9Y264 IEGR GGGSGGGSGGGS 10-His tag (C-terminus)
Oligomer
Recombinant
57 kDa (monomer)
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
85%, by SDS-PAGE under reducing conditions and visualized by silver stain.
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit