Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human APP Recombinant Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Inhibition Activity

Manufacturer:  R&D Systems™ 3466PI010

Catalog No. 3466PI010


Add to cart
This item is not returnable. View return policy

Description

Description

The Recombinant Human APP/Protease Nexin II Protein is derived from NS0. The Recombinant Human APP/Protease Nexin II Protein has been validated for the following applications: Inhibition Activity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
77 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse myeloma cell line,NS0-derived human APP protein Leu18-Leu688,with a C-terminal 6-His tag
Unconjugated
351
APP
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit