Learn More
R&D Systems™ Human Artemin Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity, Binding Activity
$459.36
Specifications
Gene ID (Entrez) | 9048 |
---|---|
Molecular Weight (g/mol) | 12 kDa (monomer) |
Name | Artemin |
Quantity | 25μg |
Source | E. coli-derived human Artemin protein Ala108-Gly220 |
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
2589AR025
|
R&D Systems™
2589AR025 |
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. |
Each for $459.36
|
|
|||||
2589AR025CF
|
R&D Systems™
2589AR025/CF |
Lyophilized from a 0.2μm filtered solution in HCl. |
Each for $459.36
|
|
|||||
Description
The Recombinant Human Artemin Protein is derived from E. coli. The Recombinant Human Artemin Protein has been validated for the following applications: Bioactivity, Binding Activity.Specifications
9048 | |
Artemin | |
E. coli-derived human Artemin protein Ala108-Gly220 | |
Disulfide-linked homodimer | |
Recombinant |
12 kDa (monomer) | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.