Learn More
R&D Systems™ Recombinant Human Azurocidin/CAP37/HBP Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Supplier: R&D Systems™ 2200SE050
This item is not returnable.
View return policy
Description
The Recombinant Human Azurocidin/CAP37/HBP Protein is derived from NS0. The Recombinant Human Azurocidin/CAP37/HBP Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in HEPES and NaCl with BSA as a carrier protein. | |
26 kDa | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
566 | |
Azurocidin/CAP37/HBP | |
Mouse myeloma cell line,NS0-derived human Azurocidin/CAP37/HBP protein Ile27-Pro250,with a C-terminal 10-His tag | |
<0.1 EU per 1μg of the protein by the LAL method. | |
Recombinant |
Provide Content Correction