Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human BAFF/BLyS/TNFSF13B (CHO-expressed) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
$407.16
Specifications
Gene ID (Entrez) | 10673 |
---|---|
Molecular Weight (g/mol) | 17.0 kDa (monomer) |
Name | BAFF/BLyS/TNFSF13B |
Quantity | 25μg |
Source | Chinese Hamster Ovary cell line,CHO-derived human BAFF/BLyS/TNFSF13B protein Ala134-Leu285 |
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
7537BF025
|
R&D Systems™
7537BF025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
Each for $407.16
|
|
|||||
7537BF025CF
|
R&D Systems™
7537BF025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
Each for $407.16
|
|
|||||
Description
The Recombinant Human BAFF/BLyS/TNFSF13B (CHO-expressed) Protein is derived from CHO. The Recombinant Human BAFF/BLyS/TNFSF13B (CHO-expressed) Protein has been validated for the following applications: Bioactivity.Specifications
10673 | |
BAFF/BLyS/TNFSF13B | |
Chinese Hamster Ovary cell line,CHO-derived human BAFF/BLyS/TNFSF13B protein Ala134-Leu285 | |
High Molecular Weight Multimer | |
Recombinant |
17.0 kDa (monomer) | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title