Learn More
R&D Systems Human Caspase-3 Recombinant Protein
Manufacturer:  R&D Systems 707C3010
 View more versions of this product
Catalog No. 707C3010
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Supplied as a 0.2μm filtered solution in PBS with BSA as a carrier protein.
17 kDa (subunit 1), 11 kDa (subunit 2)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
836
Caspase-3
10μg
Unlabeled