Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human Caspase-7 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
$545.20
Specifications
Gene ID (Entrez) | 840 |
---|---|
Molecular Weight (g/mol) | 20 kDa (p20) & 11 kDa (p11) |
Name | Caspase-7 |
Quantity | 10μg |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
823C7010CF
|
R&D Systems™
823C7010/CF |
Supplied as a 0.2μm filtered solution in HEPES, NaCl, DTT and Sucrose. |
Each of 1 for $545.20
|
|
|||||
Description
The Recombinant Human Caspase-7 Protein is derived from E. coli. The Recombinant Human Caspase-7 Protein has been validated for the following applications: Enzyme Activity.Specifications
840 | |
Caspase-7 | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. | |
Recombinant |
20 kDa (p20) & 11 kDa (p11) | |
10μg | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title