Learn More
R&D Systems Human CCL14a/HCC-1 aa 20-93 Recombinant Protein
Manufacturer:  R&D Systems 324HC010
 View more versions of this product
Catalog No. 324HC010
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
8.7 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
6358
CCL14a/HCC-1 aa 20-93
10μg
Unlabeled