Learn More
R&D Systems™ Human CCL23/Ck beta 8-1 aa 46-137 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Manufacturer:  R&D Systems™ 508CK025
 View more versions of this product
Catalog No. 508CK025
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human CCL23/Ck beta 8-1 (aa 46-137) Protein is derived from E. coli. The Recombinant Human CCL23/Ck beta 8-1 (aa 46-137) Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
10.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
E. coli-derived human CCL23/Ck beta 8-1 protein Met46-Asn137
Unconjugated
6368
CCL23/Ck beta 8-1 aa 46-137
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes