Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
R&D Systems Human CCL25/TECK Recombinant Protein
Manufacturer:  R&D Systems 334TK025
 View more versions of this product
Catalog No. 334TK025
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
14.3 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
6370
CCL25/TECK
25μg
Unlabeled