Learn More
R&D Systems™ Human CD4+ T Cell Enrichment Column, Small
Designed to prepare sub-populations of CD4 human T cells via high affinity negative selection
Manufacturer:  R&D Systems™ HCD4C1000
Catalog No. HCD4C1000
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
This item is not returnable. View return policy
Description
Description
CD4 is an approximately 55kDa type I transmembrane glycoprotein that is expressed predominantly on thymocytes and a subset of mature T cells. In humans, CD4 is also expressed to a lesser extent on monocytes and macrophage related cells.
The resulting column eluate is a highly enriched CD4+ cell subset population with minimal depletion of the selected T cells
Virtually no B cells, monocytes, or non-selected T cell subsets
Recovery of CD3+/4+ cells from all available CD3+/4+ cells ranged between 30% and 45%
Purity of recovered cells ranged between 90% and 97%, with no detectable CD3+/8+ cells among all cells recovered
Specifications
Specifications
Store the unopened product at 2 - 8° C. Do not use past expiration date.