Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CD40 Ligand/TNFSF5 (Histidine-tagged)

A New rh CD40 Ligand is Now Available! ∼10 fold better activity; HEK293 expressed; HA tag!

Supplier:  R&D Systems™ 2706CL025

 View more versions of this product

Catalog No. 2706CL025


Add to Cart
This item is not returnable. View return policy

Description

Description

Measured in a cell proliferation assay using enriched human B cells in the presence of IL-4. Spriggs, M.K. et al. (1992) J. Exp. Med. 176:1543. The ED50 for this effect is 2-12 ng/mL in the presence of 10 μg/mL of a cross-linking antibody, Mouse Anti-polyHistidine Monoclonal Antibody (Catalog # MAB050 ) and 20 ng/mL of recombinant human IL-4.
Specifications

Specifications

P29965
Lyophilized from a 0.2 µm filtered solution in HEPES, NaCl and EDTA with BSA as a carrier protein.
M.W. (Observed): 18 kDa, reducing conditions; M.W. (theoretical): 17.7 kDa (monomer)
>97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
E. coli-derived human CD40 Ligand/TNFSF5 protein Glu108-Leu261, with an N-terminal Met and 6-His tag
Homotrimer
CD40LG
Yes
CD40 Ligand/TNFSF5
Bioactivity
959
CD40 Ligand/TNFSF5 (Histidine-tagged)
25 µg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
<0.01 EU / 1 µg of the protein by the LAL method.
Unconjugated
Reconstitute at 100 µg/mL in sterile PBS containing (at least 0.1% human or bovine serum albumin.)
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit