Learn More
R&D Systems™ Recombinant Human Cystatin C Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Inhibition Activity
Supplier:  R&D Systems™ 1196PI010
Catalog No. 1196PI010
Add to Cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human Cystatin C Protein is derived from NS0. The Recombinant Human Cystatin C Protein has been validated for the following applications: Inhibition Activity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in HEPES and NaCl.
15 kDa
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
1471
Cystatin C
Mouse myeloma cell line,NS0-derived human Cystatin C protein Met1-Ala146,with a C-terminal 10-His tag
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions
Product Suggestions