Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human Dkk-2 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
$476.50
Specifications
For Use With (Application) | Bioactivity |
---|---|
Gene ID (Entrez) | 27123 |
Molecular Weight (g/mol) | 25.8 kDa |
Name | Dkk-2 |
Quantity | 10μg |
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
6628DK010CF
|
R&D Systems™
6628DK010/CF |
Lyophilized from a 0.2μm filtered solution in PBS and EDTA. |
Each for $476.50
|
|
|||||
6628DK010
|
R&D Systems™
6628DK010 |
Lyophilized from a 0.2μm filtered solution in PBS and EDTA with BSA as a carrier protein. |
Each for $476.50
|
|
|||||
Description
The Recombinant Human Dkk-2 Protein is derived from CHO. The Recombinant Human Dkk-2 Protein has been validated for the following applications: Bioactivity.Specifications
Bioactivity | |
25.8 kDa | |
10μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
27123 | |
Dkk-2 | |
Chinese Hamster Ovary cell line,CHO-derived human Dkk-2 protein Met1-Ile259 | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title