Learn More
R&D Systems™ Recombinant Human DLL1 His-tag Protein, CF Available on GSA/VA Contract for Federal Government customers only.
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Manufacturer:  R&D Systems™ 1818DL050
Catalog No. 1818DL050
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human DLL1 Protein is derived from NS0. The Recombinant Human DLL1 Protein has been validated for the following applications: Bioactivity.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
57 kDa and 43 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse myeloma cell line, NS0-derived Human DLL1 (Ser22-Gly540) & (His152-Gly540) Accession # AAG09716 DI 6-His tag
Unlabeled
28514
DLL1
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes