Learn More
R&D Systems™ Recombinant Human DLL4 His-tag Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Supplier: R&D Systems™ 1506D4050
This item is not returnable.
View return policy
Description
The Recombinant Human DLL4 Protein is derived from NS0. The Recombinant Human DLL4 Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in Tris-HCl, NaCl and PEG 3350 with BSA as a carrier protein. | |
55.6 kDa | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
54567 | |
DLL4 | |
Mouse myeloma cell line,NS0-derived human DLL4 protein Ser27-Pro524,with a C-terminal 10-His tag | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |