Learn More
R&D Systems™ Human DLL4 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Supplier:  R&D Systems™ 1506D4050/CF
Catalog No. 1506D4050CF
Add to Cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human DLL4 Protein is derived from NS0. The Recombinant Human DLL4 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS and NaCl.
55.6 kDa
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
54567
DLL4
Mouse myeloma cell line,NS0-derived human DLL4 protein Ser27-Pro524,with a C-terminal 10-His tag
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.