Learn More
R&D Systems™ Human EDA2R/TNFRSF27/XEDAR Fc Chimera Recombinant Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Manufacturer:  R&D Systems™ 1093XD050
Catalog No. 1093XD050
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human EDA2R/TNFRSF27/XEDAR Fc Chimera is derived from NS0. The Recombinant Human EDA2R/TNFRSF27/XEDAR Fc Chimera has been validated for the following applications: Binding Activity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
41.8 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse myeloma cell line,NS0-derived human EDA2R/TNFRSF27/XEDAR protein, (N-terminus) Human EDA2R (Met1-Glu136) Accession &Num; Q9HAV5 DIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Disulfide-linked homodimer
Yes
60401
EDA2R/TNFRSF27/XEDAR Fc Chimera
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated