Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human FGF-10 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
$368.88
Specifications
Gene ID (Entrez) | 2255 |
---|---|
Molecular Weight (g/mol) | Observed Molecular Wt.: 19-22 kDa, reducing conditions |
Name | FGF-10 |
Quantity | 25μg |
Source | E. coli-derived human FGF-10 protein Cys37-Ser208 & Gly41-Ser208 |
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
345FG025
|
R&D Systems™
345FG025 |
Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT with BSA as a carrier protein. |
Each for $368.88
|
|
|||||
345FG025CF
|
R&D Systems™
345FG025/CF |
Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT. |
Each for $368.88
|
|
|||||
Description
The Recombinant Human FGF-10 Protein is derived from E. coli. The Recombinant Human FGF-10 Protein has been validated for the following applications: Bioactivity.Specifications
2255 | |
FGF-10 | |
E. coli-derived human FGF-10 protein Cys37-Ser208 & Gly41-Ser208 | |
FGF10, FGF-10, fibroblast growth factor 10, Keratinocyte growth factor 2, KGF2, KGF-2, produced by fibroblasts of urinary bladder lamina propria | |
Recombinant |
Observed Molecular Wt.: 19-22 kDa, reducing conditions | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title