Learn More
R&D Systems Human Flt-3 Ligand Recombinant Protein
Manufacturer:  R&D Systems 308FK005
 View more versions of this product
Catalog No. 308FK005
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
17.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
2323
Flt-3 Ligand
5μg
Unlabeled