Learn More
R&D Systems™ Recombinant Human Follistatin 300 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Supplier: R&D Systems™ 669FO025
This item is not returnable.
View return policy
Description
The Recombinant Human Follistatin 300 Protein is derived from Sf 21 (baculovirus). The Recombinant Human Follistatin 300 Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. | |
31 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
10468 | |
Follistatin 300 aa 30-329 | |
Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Follistatin protein Gly30-Asp329,with an N-terminal Met | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |