Learn More
R&D Systems Human Follistatin 315 aa 30-344 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Manufacturer: R&D Systems 4889FN025/CF
This item is not returnable.
View return policy
Description
The Recombinant Human Follistatin (aa 30-344) Protein is derived from CHO. The Recombinant Human Follistatin (aa 30-344) Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. | |
34.7 kDa | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. | |
Chinese Hamster Ovary cell line,CHO-derived human Follistatin protein Gly30-Trp344 | |
Unconjugated |
10468 | |
Follistatin 315 aa 30-344 | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Yes |