Learn More
R&D Systems™ Human Follistatin-like 4/FSTL4 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Manufacturer:  R&D Systems™ 4890FN050
Catalog No. 4890FN050
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human Follistatin-like 4 Protein is derived from CHO. The Recombinant Human Follistatin-like 4 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
93.6 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Chinese Hamster Ovary cell line,CHO-derived human Follistatin-like 4/FSTL4 protein Trp23-Val842,with an N-terminal 7-His tag and Asn
Unconjugated
23105
Follistatin-like 4/FSTL4
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes