Learn More
R&D Systems™ Human Glypican 5 Recombinant Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
Supplier:  R&D Systems™ 2607G5050/CF
 View more versions of this product
Catalog No. 2607G5050CF
Add to Cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human Glypican 5 Protein is derived from NS0. The Recombinant Human Glypican 5 Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
59.9 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse myeloma cell line,NS0-derived human Glypican 5 protein Glu25-Thr554,with a C-terminal 6-His tag
Unconjugated
2262
Glypican 5
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes