Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human HA SUMO3 Vinyl Sulfone C-Terminal Derivative Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Supplier: R&D Systems™ UL769050
This item is not returnable.
View return policy
Description
The Recombinant Human HA-SUMO3 Vinyl Sulfone Protein is derived from E. coli. The Recombinant Human HA-SUMO3 Vinyl Sulfone Protein has been validated for the following applications: Enzyme Activity.Specifications
Xmg/mL (XμM) in 25mM MOPS pH 6.5, 150mM NaCl, 10% (v/v) Glycerol | |
Predicted MW: 12kDa | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. | |
Recombinant |
6612 | |
HA SUMO3 Vinyl Sulfone C-Terminal Derivative | |
E. coli-derived human SUMO3 protein Contains an N-terminal HA (YPYDVPDYA) tag and a C-terminal Vinyl Sulfone | |
Vinyl Sulfone | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction