Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems Human HGF R/c-MET Fc Chimera Recombinant Protein

Manufacturer:  R&D Systems 358MT100/CF

 View more versions of this product

Catalog No. 358MT100CF


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
32.5 kDa (alpha chain), 96.7 kDa (beta chain)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
4233
HGF R/c-MET Fc Chimera
100μg
Tetramer containing two disulfide-linked heterodimers (proteolytically cleaved alpha and beta subunits)
Yes
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit