Learn More
R&D Systems Human HR23A/Rad23A TUBE1 Tandem UBA (TUBE1) Agarose Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Manufacturer: R&D Systems AM125250
This item is not returnable.
View return policy
Description
The Recombinant Human HR23A Tandem UBA (TUBE1) Agarose is derived from E. coli. The Recombinant Human HR23A Tandem UBA (TUBE1) Agarose has been validated for the following applications: Enzyme Activity.Specifications
Supplied as a 50% (v/v) slurry in a solution of 20% Ethanol in deionized water | |
HR23A/Rad23A TUBE1 Tandem UBA (TUBE1) Agarose | |
E. coli-derived | |
Agarose |
5886 | |
250μL | |
12 months from date of receipt, 2 to 8° C as supplied. 3 months, 2 to 8° C under sterile conditions after opening. | |
Yes |