Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human ICAM-1/CD54 Non Allele-specific Quantikine ELISA Kit
Human ICAM-1/CD54 Non Allele-specific Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
$736.60 - $4199.20
Specifications
Assay Range | 0.63 - 40ng/mL |
---|---|
Assay Sensitivity | 0.053ng/mL |
Assay | Solid Phase Sandwich ELISA |
Conjugate | HRP |
Cross Reactivity | < 0.5% cross-reactivity observed with available related molecules. < 50% cross-species reactivity observed with species tested. |
Description
The Human ICAM-1/CD54 Non Allele-specific Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human ICAM-1/CD54 in cell culture supernates (100 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), urine (50 ul).Specifications
0.63 - 40ng/mL | |
Solid Phase Sandwich ELISA | |
< 0.5% cross-reactivity observed with available related molecules. < 50% cross-species reactivity observed with species tested. | |
Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine | |
Human | |
2.5 hr. | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |
0.053ng/mL | |
HRP | |
96-well Strip Plate | |
Natural and Recombinant Human ICAM-1 | |
ELISA | |
Cell Culture Supernates (100uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Urine (50uL) | |
BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM1, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title