Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human IFN-gamma R1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
$518.52
Specifications
Gene ID (Entrez) | 3459 |
---|---|
Molecular Weight (g/mol) | 25 kDa |
Name | IFN-gamma R1/CD119 |
Quantity | 100μg |
Source | Mouse myeloma cell line,NS0-derived human IFN-gamma R1/CD119 protein Met1-Gly245 |
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
673IR100
|
R&D Systems™
673IR100 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
Each for $518.52
|
|
|||||
673IR100CF
|
R&D Systems™
673IR100/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
Each for $518.52
|
|
|||||
Description
The Recombinant Human IFN-gamma R1 Protein is derived from NS0. The Recombinant Human IFN-gamma R1 Protein has been validated for the following applications: Bioactivity.Specifications
3459 | |
IFN-gamma R1/CD119 | |
Mouse myeloma cell line,NS0-derived human IFN-gamma R1/CD119 protein Met1-Gly245 | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
25 kDa | |
100μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title