Learn More
R&D Systems Human IL-15 Recombinant Protein
Manufacturer:  R&D Systems 247IL005/CF
 View more versions of this product
Catalog No. 247IL005CF
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
12.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
3600
IL-15
5μg
Unlabeled