Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
R&D Systems Human IL-15 Recombinant Protein
Manufacturer:  R&D Systems 247IL025/CF
 View more versions of this product
Catalog No. 247IL025CF
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
12.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
3600
IL-15
25μg
Unlabeled