Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human IL-17E Protein
A New and Improved rh IL-17E is Now Available! The new protein is mammalian cell expressed!
$459.36
Specifications
Gene ID (Entrez) | 64806 |
---|---|
Name | IL-17E/IL-25 |
Structural Form | Disulfide-linked homodimer |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
1258IL025
|
R&D Systems™
1258IL025 |
Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
Each for $459.36
|
|
|||||
1258IL025CF
|
R&D Systems™
1258IL025/CF |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
Each for $459.36
|
|
|||||
Description
Measured by its ability to induce CXCL1/GRO alpha secretion in HT-29 human colon adenocarcinoma cells. The ED50 for this effect is 0.25-1.5 ng/mL.Specifications
64806 | |
Disulfide-linked homodimer | |
Recombinant |
IL-17E/IL-25 | |
Unconjugated |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title