Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human IL-17E (Human Cell-expressed) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
$459.36
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. |
---|---|
Gene ID (Entrez) | 64806 |
Molecular Weight (g/mol) | 17 kDa (monomer) |
Name | IL-17E/IL-25 |
Quantity | 25μg |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
8134IL025
|
R&D Systems™
8134IL025 |
25μg |
Each of 1 for $459.36
|
|
|||||
Description
The Recombinant Human IL-17E (Human Cell-expressed) Protein is derived from HEK293. The Recombinant Human IL-17E (Human Cell-expressed) Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. | |
17 kDa (monomer) | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE with silver staining. |
64806 | |
IL-17E/IL-25 | |
Human embryonic kidney cell,HEK293-derived human IL-17E/IL-25 protein Tyr33-Gly177 | |
Disulfide-linked homodimer | |
Recombinant |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title