Learn More
R&D Systems™ Human IL-1F7/FIL1 zeta Recombinant Protein Available on GSA/VA Contract for Federal Government customers only.
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
Manufacturer:  R&D Systems™ 1975IL025
Catalog No. 1975IL025
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human IL-37/IL-1F7 Protein is derived from E. coli. The Recombinant Human IL-37/IL-1F7 Protein has been validated for the following applications: Binding Activity.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS and DTT.
18.6 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
E. coli-derived human IL-37/IL-1F7 protein Lys27-Asp192,with an N-terminal Met
Unconjugated
27178
IL-1F7/FIL1 zeta
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes