R&D Systems Human IL-36 beta/IL-1F8 (aa 5-157) Recombinant Protein

For use in cell or tissue culture applications.

Manufacturer: R&D Systems 6834IL010

 View more versions of this product

Catalog No. 6834IL010

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Molecular Weight 17.2 kDa
    Name IL-36 beta/IL-1F8 (aa 5-157)
    Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
    Quantity 10μg
    Recombinant Yes
    Source Human
    Conjugate Unlabeled
    Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
    Gene ID (Entrez) 27177

    • Source: E. coli-derived, Arg5-Glu157
    • Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
    • Molecular Weight: 17.2 kDa
    • Formulation: Lyophilized from a 0.2µm filtered solution in PBS.
    • Entrez Gene ID: 27177
    • Regulatory Status: RUO

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit