Learn More
R&D Systems Human IL-6 Recombinant Protein
Manufacturer:  R&D Systems 206IL010/CF
 View more versions of this product
Catalog No. 206IL010CF
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS and NaCl.
20.3 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
3569
IL-6
10μg
Unlabeled