Learn More
R&D Systems Human IL-9 Recombinant Protein
Manufacturer:  R&D Systems 209IL010
 View more versions of this product
Catalog No. 209IL010
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
14.1 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
3578
IL-9
10μg
Unlabeled