R&D Systems Human LR3 IGF-I Recombinant Protein

Manufacturer: R&D Systems 8335G101M

 View more versions of this product

Catalog No. 83-35G-101M

  Check Availability
Add to Cart

    Specifications

    Accession Number P05019
    Molecular Weight 9kDa
    Name LR3 IGF-I
    Quantity 1mg
    Shelf Life 365 days
    Purity 95%, by SDS-PAGE with silver staining
    Recombinant Yes
    Source Human
    Conjugate Unlabeled
    Formulation Lyophilized from a 0.2μm filtered solution in phosphate buffer, pH 7.2
    Gene ID (Entrez) 3479
    Preparation Method Reconstitute at 500μg/mL in sterile PBS.
    Quality Control Testing SDS-PAGE: 7kDa, reducing conditions
    Storage Requirements -20 to -70°C
    Regulatory Status RUO
    Without Additives Carrier Protein
    Termination Met-Phe-Pro-Ala-Met-Pro-Leu-Ser-Ser-Leu

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit